Assay Details
Gene Symbol: AGL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037046.2 | NP_001032123.1 | 7 - 8 | 834 | 122 | |
NM_001203665.2 | NP_001190594.1 | 6 - 7 | 685 | 122 | ||
NM_001345529.1 | NP_001330224.1 | 6 - 7 | 953 | 226 | ||
NM_001345530.1 | NP_001330225.1 | 6 - 7 | 941 | 226 | ||
NM_001345531.1 | NP_001330226.1 | 6 - 7 | 1101 | 122 | ||
NM_001345532.1 | NP_001330218.1 | 4 - 5 | 1112 | 226 | ||
NM_001345533.1 | NP_001330219.1 | 6 - 7 | 753 | 122 | ||
NM_001345535.1 | NP_001330221.1 | 5 - 6 | 697 | 226 | ||
NM_001345536.1 | NP_001330228.1 | 4 - 5 | 1309 | 226 | ||
NM_001345537.1 | NP_001330229.1 | 5 - 6 | 1231 | 226 | ||
NM_001345538.1 | NP_001330220.1 | 5 - 6 | 1219 | 226 | ||
NM_001345539.1 | NP_001330227.1 | 6 - 7 | 1115 | 122 | ||
NM_125610.4 | NP_568952.1 | 7 - 8 | 848 | 122 | ||
NM_180916.1 | NP_851247.1 | 6 - 7 | 884 | 122 | ||
GenBank mRNA | AY054220.1 | - | 6 - 7 | 885 | 122 | |
AY065206.1 | - | 6 - 7 | 885 | 122 | ||
AY066035.1 | - | 6 - 7 | 509 | 122 | ||
AY096509.1 | - | 6 - 7 | 509 | 122 | ||
AY141213.1 | - | 6 - 7 | 509 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
