Assay Details
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 9 - 10 | 1186 | 88 | |
GenBank mRNA | AB017643.1 | - | 9 - 10 | 892 | 88 | |
AY087793.1 | - | 9 - 10 | 911 | 88 | ||
AY094441.1 | - | 9 - 10 | 1008 | 88 | ||
AY125536.1 | - | 9 - 10 | 853 | 88 | ||
U72505.1 | - | 9 - 10 | 878 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
