Assay Details
Gene Symbol: UPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035773.2 | NP_001030850.1 | 6 - 7 | 2327 | 92 | |
NM_115169.3 | NP_190877.1 | 6 - 7 | 2330 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
