Assay Details
Gene Symbol: AT3G53100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115170.5 | NP_190878.2 | 2 - 3 | 816 | 86 | |
GenBank mRNA | AK229079.1 | - | 2 - 3 | 414 | 86 | |
BT033018.1 | - | 2 - 3 | 374 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
