Assay Details
Gene Symbol: AT3G53100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115170.5 | NP_190878.2 | 4 - 5 | 1307 | 68 | |
GenBank mRNA | AK229079.1 | - | 4 - 5 | 905 | 68 | |
BT033018.1 | - | 4 - 5 | 865 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
