Assay Details
Gene Symbol: GCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339630.1 | NP_001325627.1 | 6 - 7 | 1254 | 99 | |
NM_001339631.1 | NP_001325626.1 | 6 - 7 | 1252 | 99 | ||
NM_115236.4 | NP_190944.2 | 6 - 7 | 1343 | 99 | ||
GenBank mRNA | BT012227.1 | - | 6 - 7 | 1193 | 99 | |
BT012412.1 | - | 6 - 7 | 1095 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
