Assay Details
Gene Symbol: PMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115349.6 | NP_191052.2 | 2 - 3 | 1571 | 75 | |
GenBank mRNA | AF534079.1 | - | 2 - 3 | 1220 | 75 | |
AY058870.1 | - | 2 - 3 | 1257 | 75 | ||
AY074331.1 | - | 2 - 3 | 1253 | 75 | ||
AY096739.1 | - | 2 - 3 | 1045 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
