Assay Details
Gene Symbol: AT3G55920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115450.4 | NP_567029.1 | 3 - 4 | 327 | 133 | |
GenBank mRNA | AY088764.1 | - | 3 - 4 | 248 | 133 | |
AY568519.1 | - | 3 - 4 | 223 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
