Assay Details
Gene Symbol: AT3G55920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115450.4 | NP_567029.1 | 4 - 5 | 371 | 78 | |
GenBank mRNA | AY088764.1 | - | 4 - 5 | 292 | 78 | |
AY568519.1 | - | 4 - 5 | 267 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
