Assay Details
Gene Symbol: EDA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339835.1 | NP_001327450.1 | 14 - 15 | 2031 | 61 | |
NM_115558.6 | NP_191258.2 | 15 - 16 | 1931 | 61 | ||
GenBank mRNA | AY069919.1 | - | 15 - 16 | 1818 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
