Assay Details
Gene Symbol: AT3G57220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115582.3 | NP_191281.1 | 7 - 8 | 1117 | 97 | |
GenBank mRNA | AK175168.1 | - | 7 - 8 | 1057 | 97 | |
AY088931.1 | - | 7 - 8 | 1053 | 97 | ||
BT029005.1 | - | 7 - 8 | 996 | 97 | ||
KY906095.1 | - | 7 - 8 | 996 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
