Assay Details
Gene Symbol: ADSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115621.5 | NP_191320.1 | 1 - 2 | 414 | 106 | |
GenBank mRNA | AY054606.1 | - | 1 - 2 | 352 | 106 | |
AY081461.1 | - | 1 - 2 | 297 | 106 | ||
U49389.1 | - | 1 - 2 | 330 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
