Assay Details
Gene Symbol: ADSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115621.5 | NP_191320.1 | 2 - 3 | 1111 | 87 | |
GenBank mRNA | AY054606.1 | - | 2 - 3 | 1049 | 87 | |
AY081461.1 | - | 2 - 3 | 994 | 87 | ||
U49389.1 | - | 2 - 3 | 1027 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
