Assay Details
Gene Symbol: DEG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123346.5 | NP_568575.1 | 11 - 12 | 1227 | 109 | |
NM_203134.2 | NP_974863.1 | 11 - 12 | 1152 | 109 | ||
GenBank mRNA | AY056381.1 | - | 11 - 12 | 1171 | 109 | |
AY090319.1 | - | 11 - 12 | 1108 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
