Assay Details
Gene Symbol: DEG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123346.5 | NP_568575.1 | 4 - 5 | 564 | 66 | |
NM_203134.2 | NP_974863.1 | 4 - 5 | 531 | 66 | ||
GenBank mRNA | AY056381.1 | - | 4 - 5 | 508 | 66 | |
AY090319.1 | - | 4 - 5 | 445 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
