Assay Details
Gene Symbol: SAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198200.2 | NP_001185129.1 | 7 - 8 | 1173 | 98 | |
NM_001333042.1 | NP_001319133.1 | 7 - 8 | 1161 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
