Assay Details
Gene Symbol: AT1G48170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103713.3 | NP_564522.1 | 2 - 3 | 292 | 59 | |
GenBank mRNA | AK228677.1 | - | 2 - 3 | 269 | 59 | |
AY070372.1 | - | 2 - 3 | 268 | 59 | ||
AY086896.1 | - | 2 - 3 | 269 | 59 | ||
AY133765.1 | - | 2 - 3 | 245 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
