Assay Details
Gene Symbol: AT1G48170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103713.3 | NP_564522.1 | 3 - 4 | 367 | 80 | |
GenBank mRNA | AK228677.1 | - | 3 - 4 | 344 | 80 | |
AY070372.1 | - | 3 - 4 | 343 | 80 | ||
AY086896.1 | - | 3 - 4 | 344 | 80 | ||
AY133765.1 | - | 3 - 4 | 320 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
