Assay Details
Gene Symbol: PDAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121367.3 | NP_196868.1 | 1 - 2 | 1039 | 119 | |
GenBank mRNA | AY052715.1 | - | 1 - 2 | 506 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
