Assay Details
Gene Symbol: AT5G17360 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121742.2 | NP_197238.1 | 1 | 1212 | 97 | |
GenBank mRNA | AK118392.1 | - | 1 | 1212 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
