Assay Details
Gene Symbol: AT5G17380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121744.4 | NP_197240.1 | 1 - 2 | 1592 | 100 | |
GenBank mRNA | AY099774.1 | - | 1 - 2 | 1426 | 100 | |
BT000284.1 | - | 1 - 2 | 1400 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
