Assay Details
Gene Symbol: AT5G18140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121819.6 | NP_197315.1 | 1 - 2 | 418 | 85 | |
GenBank mRNA | AY093186.1 | - | 1 - 2 | 334 | 85 | |
BT003386.1 | - | 1 - 2 | 310 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
