Assay Details
Gene Symbol: ACL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085136.1 | NP_001078605.1 | 3 - 4 | 380 | 96 | |
NM_121958.4 | NP_568376.1 | 2 - 3 | 440 | 96 | ||
GenBank mRNA | AF184094.1 | - | 2 - 3 | 246 | 96 | |
AY087927.1 | - | 2 - 3 | 310 | 96 | ||
AY127003.1 | - | 2 - 3 | 315 | 96 | ||
BT002254.1 | - | 2 - 3 | 226 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
