Assay Details
Gene Symbol: ACL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085136.1 | NP_001078605.1 | 9 - 10 | 1092 | 101 | |
NM_121958.4 | NP_568376.1 | 8 - 9 | 1152 | 101 | ||
GenBank mRNA | AF184094.1 | - | 8 - 9 | 958 | 101 | |
AY127003.1 | - | 8 - 9 | 1027 | 101 | ||
BT002254.1 | - | 8 - 9 | 938 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
