Assay Details
Gene Symbol: KAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202728.1 | NP_001189657.1 | 4 - 5 | 1214 | 91 | |
NM_128803.4 | NP_180803.1 | 3 - 4 | 1072 | 91 | ||
GenBank mRNA | BT029168.1 | - | 3 - 4 | 974 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
