Assay Details
Gene Symbol: ROC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161212.1 | NP_001154684.1 | 3 - 4 | 213 | 112 | |
NM_001203230.1 | NP_001190159.1 | 1 - 2 | 198 | 112 | ||
NM_116068.5 | NP_191762.1 | 1 - 2 | 196 | 112 | ||
GenBank mRNA | AY059843.1 | - | 1 - 2 | 117 | 112 | |
AY093284.1 | - | 1 - 2 | 54 | 112 | ||
L14845.1 | - | 1 - 2 | 81 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
