Assay Details
Gene Symbol: EDA32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116086.3 | NP_191780.1 | 1 - 2 | 499 | 120 | |
GenBank mRNA | AY735630.1 | - | 1 - 2 | 470 | 120 | |
AY924806.1 | - | 1 - 2 | 277 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
