Assay Details
Gene Symbol: PLDBETA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340237.1 | NP_001329750.1 | 2 - 3 | 1368 | 74 | |
NM_001340238.1 | NP_001329749.1 | 2 - 3 | 1191 | 74 | ||
NM_116245.2 | NP_567160.1 | 1 - 2 | 1473 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
