Assay Details
Gene Symbol: PLDBETA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340237.1 | NP_001329750.1 | 6 - 7 | 2000 | 112 | |
NM_001340238.1 | NP_001329749.1 | 6 - 7 | 1823 | 112 | ||
NM_116245.2 | NP_567160.1 | 5 - 6 | 2105 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
