Assay Details
Gene Symbol: EDA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036486.3 | NP_001031563.1 | 2 - 3 | 330 | 93 | |
NM_116253.3 | NP_567168.1 | 1 - 2 | 435 | 93 | ||
GenBank mRNA | AK317127.1 | - | 2 - 3 | 329 | 93 | |
AY049241.1 | - | 1 - 2 | 435 | 93 | ||
AY090270.1 | - | 1 - 2 | 221 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
