Assay Details
Gene Symbol: EDA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036486.3 | NP_001031563.1 | 3 - 4 | 401 | 108 | |
NM_116253.3 | NP_567168.1 | 2 - 3 | 506 | 108 | ||
GenBank mRNA | AK317127.1 | - | 3 - 4 | 400 | 108 | |
AY049241.1 | - | 2 - 3 | 506 | 108 | ||
AY090270.1 | - | 2 - 3 | 292 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
