Assay Details
Gene Symbol: LysoPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104154.4 | NP_175685.1 | 1 - 2 | 543 | 68 | |
GenBank mRNA | AY054577.1 | - | 1 - 2 | 371 | 68 | |
BT008729.1 | - | 1 - 2 | 337 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
