Assay Details
Gene Symbol: AT1G55890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104466.4 | NP_175985.1 | 1 | 1479 | 117 | |
GenBank mRNA | AY074876.1 | - | 1 | 1376 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
