Assay Details
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333872.1 | NP_001320772.1 | 4 - 5 | 846 | 90 | |
NM_001333873.1 | NP_001320771.1 | 3 - 4 | 895 | 90 | ||
NM_104690.4 | NP_176206.2 | 4 - 5 | 846 | 90 | ||
GenBank mRNA | AK118901.1 | - | 4 - 5 | 742 | 90 | |
BT005502.1 | - | 4 - 5 | 466 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
