Assay Details
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333872.1 | NP_001320772.1 | 5 - 6 | 946 | 98 | |
NM_001333873.1 | NP_001320771.1 | 4 - 5 | 995 | 98 | ||
NM_104690.4 | NP_176206.2 | 5 - 6 | 946 | 98 | ||
GenBank mRNA | AK118901.1 | - | 5 - 6 | 842 | 98 | |
BT005502.1 | - | 5 - 6 | 566 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
