Assay Details
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333872.1 | NP_001320772.1 | 7 - 8 | 1093 | 98 | |
NM_001333873.1 | NP_001320771.1 | 6 - 7 | 1142 | 98 | ||
NM_104690.4 | NP_176206.2 | 7 - 8 | 1093 | 98 | ||
GenBank mRNA | AK118901.1 | - | 7 - 8 | 989 | 98 | |
BT005502.1 | - | 7 - 8 | 713 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
