Assay Details
Gene Symbol: ARL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333872.1 | NP_001320772.1 | 8 - 9 | 1203 | 121 | |
NM_001333873.1 | NP_001320771.1 | 7 - 8 | 1252 | 121 | ||
NM_104690.4 | NP_176206.2 | 8 - 9 | 1203 | 121 | ||
GenBank mRNA | AK118901.1 | - | 8 - 9 | 1099 | 121 | |
BT005502.1 | - | 8 - 9 | 823 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
