Assay Details
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084274.2 | NP_001077743.1 | 4 - 5 | 694 | 85 | |
NM_001333892.1 | NP_001319276.1 | 4 - 5 | 707 | 85 | ||
NM_104730.3 | NP_564757.1 | 4 - 5 | 694 | 85 | ||
GenBank mRNA | BT025737.1 | - | 2 - 3 | 383 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
