Assay Details
Gene Symbol: EDA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084535.1 | NP_001078004.1 | 1 - 2 | 78 | 62 | |
NM_129039.4 | NP_181032.1 | 1 - 2 | 195 | 62 | ||
GenBank mRNA | AY099664.1 | - | 1 - 2 | 70 | 62 | |
AY128851.1 | - | 1 - 2 | 58 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
