Assay Details
Gene Symbol: EDA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129039.4 | NP_181032.1 | 5 - 6 | 707 | 72 | |
GenBank mRNA | AY099664.1 | - | 5 - 6 | 582 | 72 | |
AY128851.1 | - | 5 - 6 | 570 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
