Assay Details
Gene Symbol: EDA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336534.1 | NP_001324447.1 | 2 - 3 | 1288 | 75 | |
NM_129045.2 | NP_181038.2 | 2 - 3 | 1189 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
