Assay Details
Gene Symbol: NET1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340967.1 | NP_001329579.1 | 4 - 5 | 4431 | 59 | |
NM_001340968.1 | NP_001329578.1 | 4 - 5 | 4311 | 59 | ||
NM_001340969.1 | NP_001329577.1 | 3 - 4 | 4283 | 59 | ||
NM_117561.6 | NP_193212.4 | 3 - 4 | 4526 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
