Assay Details
Gene Symbol: MES16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117770.5 | NP_193402.1 | 1 - 2 | 608 | 89 | |
GenBank mRNA | AY088316.1 | - | 1 - 2 | 427 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
