Assay Details
Gene Symbol: MYC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117897.4 | NP_193522.1 | 1 | 2217 | 127 | |
GenBank mRNA | AK221507.1 | - | 1 | 2111 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
