Assay Details
Gene Symbol: AT1G71250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105794.4 | NP_177281.1 | 2 - 3 | 419 | 83 | |
GenBank mRNA | AK117368.1 | - | 2 - 3 | 411 | 83 | |
BT005325.1 | - | 2 - 3 | 404 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
