Assay Details
Gene Symbol: AT4G23820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118513.3 | NP_194113.1 | 1 - 2 | 503 | 113 | |
GenBank mRNA | AY062697.1 | - | 1 - 2 | 363 | 113 | |
AY128793.1 | - | 1 - 2 | 322 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
