Assay Details
Gene Symbol: CR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126439.4 | NP_178487.1 | 15 - 16 | 2005 | 122 | |
NM_179601.1 | NP_849932.1 | 15 - 16 | 1850 | 122 | ||
GenBank mRNA | AF436826.1 | - | 15 - 16 | 1859 | 122 | |
AX765864.1 | - | 15 - 16 | 1776 | 122 | ||
AY053403.1 | - | 15 - 16 | 1844 | 122 | ||
BT002234.1 | - | 15 - 16 | 1776 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
