Assay Details
Gene Symbol: CR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126439.4 | NP_178487.1 | 18 - 19 | 2380 | 99 | |
NM_179601.1 | NP_849932.1 | 18 - 19 | 2225 | 99 | ||
GenBank mRNA | AF436826.1 | - | 18 - 19 | 2234 | 99 | |
AX765864.1 | - | 18 - 19 | 2151 | 99 | ||
AY053403.1 | - | 18 - 19 | 2219 | 99 | ||
BT002234.1 | - | 18 - 19 | 2151 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
