Assay Details
Gene Symbol: CR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126439.4 | NP_178487.1 | 1 - 2 | 474 | 94 | |
NM_179601.1 | NP_849932.1 | 1 - 2 | 328 | 94 | ||
GenBank mRNA | AF436826.1 | - | 1 - 2 | 328 | 94 | |
AX765864.1 | - | 1 - 2 | 245 | 94 | ||
AY053403.1 | - | 1 - 2 | 313 | 94 | ||
BT002234.1 | - | 1 - 2 | 245 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
