Assay Details
Gene Symbol: NUDT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335236.1 | NP_001324657.1 | 5 - 6 | 861 | 133 | |
NM_126476.3 | NP_178524.2 | 4 - 5 | 539 | 133 | ||
GenBank mRNA | BT010760.1 | - | 4 - 5 | 539 | 133 | |
BT011326.1 | - | 4 - 5 | 496 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
